Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128315.4 | NP_180324.1 | 2 - 3 | 492 | 68 | |
GenBank mRNA | AB024715.1 | - | 2 - 3 | 282 | 68 | |
AY065211.1 | - | 2 - 3 | 426 | 68 | ||
AY096515.1 | - | 2 - 3 | 262 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |