Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCPL51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036357.1 | NP_001031434.1 | 1 - 2 | 124 | 108 | |
NM_128351.3 | NP_565663.1 | 1 - 2 | 270 | 108 | ||
GenBank mRNA | AF412049.1 | - | 1 - 2 | 207 | 108 | |
BT003041.1 | - | 1 - 2 | 137 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |