Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BLH8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128358.3 | NP_180366.1 | 3 - 4 | 1531 | 84 | |
GenBank mRNA | AK119153.1 | - | 3 - 4 | 1481 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |