Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNGC15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336157.1 | NP_001324840.1 | 2 - 3 | 905 | 73 | |
NM_128386.2 | NP_180393.1 | 3 - 4 | 1305 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |