Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128524.4 | NP_565687.1 | 1 | 817 | 141 | |
GenBank mRNA | AY050344.1 | - | 1 | 521 | 141 | |
AY084905.1 | - | 1 | 549 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |