Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MKK5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338511.1 | NP_001319606.1 | 1 | 1504 | 147 | |
NM_113017.4 | NP_188759.1 | 1 | 1377 | 147 | ||
GenBank mRNA | AB015316.1 | - | 1 | 1232 | 147 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |