Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARFC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203022.1 | NP_001189951.1 | 3 - 4 | 424 | 74 | |
NM_113195.4 | NP_188935.1 | 3 - 4 | 441 | 74 | ||
GenBank mRNA | AY063958.1 | - | 2 - 3 | 350 | 74 | |
AY086337.1 | - | 2 - 3 | 368 | 74 | ||
AY096401.1 | - | 1 - 2 | 186 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |