Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RFR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_113262.4 | NP_189001.1 | 3 - 4 | 966 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |