Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOPII | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001125214.2 | NP_001118686.1 | 13 - 14 | 3308 | 139 | |
NM_113294.3 | NP_189031.1 | 13 - 14 | 3318 | 139 | ||
GenBank mRNA | L21015.1 | - | 13 - 14 | 3204 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |