Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MSH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338677.1 | NP_001326989.1 | 8 - 9 | 1125 | 89 | |
NM_113339.4 | NP_189075.2 | 13 - 14 | 1418 | 89 | ||
GenBank mRNA | AY191303.1 | - | 13 - 14 | 1384 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |