Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338755.1 | NP_001319643.1 | 5 - 6 | 1219 | 67 | |
NM_001338756.1 | NP_001327930.1 | 5 - 6 | 1304 | 67 | ||
NM_113450.5 | NP_566769.1 | 6 - 7 | 1223 | 67 | ||
GenBank mRNA | AF198179.1 | - | 5 - 6 | 948 | 67 | |
AF360152.1 | - | 5 - 6 | 1054 | 67 | ||
AY142597.1 | - | 5 - 6 | 898 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |