Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_113576.4 | NP_566796.2 | 4 - 5 | 1052 | 83 | |
GenBank mRNA | AF428431.1 | - | 4 - 5 | 735 | 83 | |
AY058107.1 | - | 4 - 5 | 729 | 83 | ||
AY058140.1 | - | 4 - 5 | 735 | 83 | ||
AY075637.1 | - | 4 - 5 | 737 | 83 | ||
AY142053.1 | - | 4 - 5 | 688 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |