Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DXR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203671.1 | NP_001190600.1 | 2 - 3 | 415 | 108 | |
NM_125674.3 | NP_201085.1 | 2 - 3 | 463 | 108 | ||
GenBank mRNA | AF148852.1 | - | 2 - 3 | 371 | 108 | |
AY045634.1 | - | 2 - 3 | 410 | 108 | ||
AY050802.1 | - | 2 - 3 | 409 | 108 | ||
AY054682.1 | - | 2 - 3 | 371 | 108 | ||
AY081453.1 | - | 2 - 3 | 286 | 108 | ||
AY084775.1 | - | 2 - 3 | 371 | 108 | ||
AY091405.1 | - | 2 - 3 | 286 | 108 | ||
AY098952.1 | - | 2 - 3 | 286 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |