Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FLS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345596.1 | NP_001332248.1 | 1 - 2 | 537 | 142 | |
NM_125753.3 | NP_201163.1 | 1 - 2 | 537 | 142 | ||
GenBank mRNA | BT031358.1 | - | 1 - 2 | 385 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |