Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RABH1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203687.1 | NP_001190616.1 | 4 - 5 | 450 | 101 | |
NM_125898.2 | NP_201304.1 | 4 - 5 | 503 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |