Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PUB38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125917.3 | NP_201323.1 | 1 | 1761 | 62 | |
GenBank mRNA | BT015352.1 | - | 1 | 1777 | 62 | |
BT015720.1 | - | 1 | 1558 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |