Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PP2-A12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332044.1 | NP_001322372.1 | 1 - 2 | 550 | 117 | |
NM_101141.4 | NP_172731.1 | 1 - 2 | 620 | 117 | ||
GenBank mRNA | AK117662.1 | - | 1 - 2 | 485 | 117 | |
BT005109.1 | - | 1 - 2 | 328 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |