Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RSH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101166.4 | NP_172755.1 | 3 - 4 | 1135 | 68 | |
GenBank mRNA | BT023437.1 | - | 3 - 4 | 1006 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |