Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JAC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_106166.4 | NP_565101.1 | 1 - 2 | 706 | 71 | |
GenBank mRNA | AB158477.1 | - | 1 - 2 | 420 | 71 | |
AY057504.1 | - | 1 - 2 | 438 | 71 | ||
AY103303.1 | - | 1 - 2 | 407 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |