Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_106258.4 | NP_177736.1 | 1 | 1201 | 141 | |
GenBank mRNA | AF375397.1 | - | 1 | 1091 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |