Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EFS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084367.2 | NP_001077836.1 | 9 - 10 | 3481 | 157 | |
NM_106379.9 | NP_177854.6 | 9 - 10 | 3633 | 157 | ||
GenBank mRNA | EU014690.1 | - | 8 - 9 | 3346 | 157 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |