Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LACS9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001334788.1 | NP_001321320.1 | 3 - 4 | 1131 | 96 | |
NM_106407.7 | NP_177882.1 | 4 - 5 | 1058 | 96 | ||
GenBank mRNA | AF503759.1 | - | 3 - 4 | 783 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |