Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LACS9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001334788.1 | NP_001321320.1 | 5 - 6 | 1443 | 87 | |
NM_106407.7 | NP_177882.1 | 6 - 7 | 1370 | 87 | ||
GenBank mRNA | AF503759.1 | - | 5 - 6 | 1095 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |