Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PUM7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001334820.1 | NP_001322141.1 | 5 - 6 | 1998 | 69 | |
NM_106466.2 | NP_177940.2 | 4 - 5 | 1852 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |