Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001334832.1 | NP_001319403.1 | 17 - 18 | 4322 | 79 | |
NM_001334833.1 | NP_001322931.1 | 16 - 17 | 4281 | 79 | ||
NM_001334834.1 | NP_001322932.1 | 16 - 17 | 4248 | 79 | ||
NM_106505.5 | NP_177979.1 | 16 - 17 | 2512 | 79 | ||
GenBank mRNA | KJ138926.1 | - | 15 - 16 | 2306 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |