Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: J8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_106740.5 | NP_178207.1 | 1 - 2 | 412 | 71 | |
GenBank mRNA | AF332461.1 | - | 1 - 2 | 248 | 71 | |
AY040017.1 | - | 1 - 2 | 384 | 71 | ||
AY079369.1 | - | 1 - 2 | 248 | 71 | ||
AY088122.1 | - | 1 - 2 | 385 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |