Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAC7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337807.1 | NP_001326525.1 | 4 - 5 | 2123 | 83 | |
NM_111756.3 | NP_187533.1 | 5 - 6 | 1748 | 83 | ||
GenBank mRNA | AK117639.1 | - | 5 - 6 | 1635 | 83 | |
BT004971.1 | - | 5 - 6 | 1570 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |