Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIP5K9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035589.3 | NP_001030666.1 | 6 - 7 | 2373 | 81 | |
NM_001202923.1 | NP_001189852.1 | 6 - 7 | 2303 | 81 | ||
NM_001337850.1 | NP_001327647.1 | 6 - 7 | 2317 | 81 | ||
NM_111827.3 | NP_187603.1 | 5 - 6 | 3169 | 81 | ||
GenBank mRNA | AJ810853.1 | - | 5 - 6 | 1903 | 81 | |
AK229815.1 | - | 6 - 7 | 2152 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |