Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LYC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084662.1 | NP_001078131.1 | 2 | 1128 | 101 | |
NM_111858.2 | NP_187634.1 | 1 | 1342 | 101 | ||
GenBank mRNA | AY059749.1 | - | 1 | 1342 | 101 | |
AY091396.1 | - | 1 | 1149 | 101 | ||
U50739.1 | - | 1 | 1322 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |