Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111872.6 | NP_187648.1 | 4 - 5 | 1499 | 69 | |
GenBank mRNA | AY063863.1 | - | 4 - 5 | 1424 | 69 | |
AY096363.1 | - | 4 - 5 | 1336 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |