Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337916.1 | NP_001319522.1 | 6 - 7 | 1003 | 112 | |
NM_111957.5 | NP_566388.2 | 6 - 7 | 905 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |