Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CARA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084749.1 | NP_001078218.1 | 4 - 5 | 471 | 104 | |
NM_113690.3 | NP_566824.1 | 6 - 7 | 756 | 104 | ||
GenBank mRNA | AY046004.1 | - | 6 - 7 | 687 | 104 | |
AY079315.1 | - | 6 - 7 | 613 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |