Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MDAR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_113698.4 | NP_189420.1 | 3 - 4 | 488 | 74 | |
GenBank mRNA | AY039980.1 | - | 3 - 4 | 346 | 74 | |
AY133800.1 | - | 3 - 4 | 249 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |