Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL12-A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_113699.3 | NP_189421.1 | 1 | 900 | 103 | |
GenBank mRNA | AY056124.1 | - | 1 | 850 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |