Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UVH3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338928.1 | NP_001319661.1 | 15 - 16 | 3675 | 151 | |
NM_001338929.1 | NP_001325749.1 | 15 - 16 | 3675 | 151 | ||
NM_001338930.1 | NP_001325748.1 | 14 - 15 | 3679 | 151 | ||
NM_001338931.1 | NP_001325750.1 | 12 - 13 | 3129 | 151 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |