Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AT3G29070 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_113829.3 | NP_189550.2 | 1 - 2 | 359 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |