Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHOT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035737.2 | NP_001030814.1 | 19 - 20 | 3100 | 107 | |
NM_114447.4 | NP_190164.1 | 20 - 21 | 3098 | 107 | ||
GenBank mRNA | AF030864.1 | - | 20 - 21 | 2953 | 107 | |
AF360218.1 | - | 19 - 20 | 2907 | 107 | ||
AY040062.1 | - | 19 - 20 | 2890 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |