Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HRS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_114479.5 | NP_190196.1 | 6 - 7 | 838 | 96 | |
GenBank mRNA | AF020715.1 | - | 6 - 7 | 774 | 96 | |
AY056233.1 | - | 6 - 7 | 779 | 96 | ||
AY117252.1 | - | 6 - 7 | 769 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |