Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MEE39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339245.1 | NP_001326106.1 | 1 - 2 | 270 | 94 | |
NM_114500.5 | NP_190217.2 | 1 - 2 | 270 | 94 | ||
GenBank mRNA | FJ708732.1 | - | 1 - 2 | 85 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |