Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203097.2 | NP_001190026.1 | 11 - 12 | 1918 | 141 | |
NM_114607.3 | NP_190323.1 | 11 - 12 | 1927 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |