Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLDH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_114662.3 | NP_190376.1 | 5 - 6 | 1844 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |