Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SR34a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084795.1 | NP_001078264.1 | 2 - 3 | 281 | 91 | |
NM_001203112.1 | NP_001190041.1 | 3 - 4 | 533 | 91 | ||
NM_001339417.1 | NP_001325875.1 | 1 - 2 | 1247 | 91 | ||
NM_001339418.1 | NP_001325874.1 | 2 - 3 | 1077 | 91 | ||
NM_001339419.1 | NP_001325876.1 | 2 - 3 | 712 | 91 | ||
NM_001339420.1 | NP_001325877.1 | 3 - 4 | 505 | 91 | ||
NM_114803.6 | NP_190512.3 | 2 - 3 | 318 | 91 | ||
GenBank mRNA | BT030067.1 | - | 1 - 2 | 88 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |