Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARLA1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339443.1 | NP_001327086.1 | 5 - 6 | 1038 | 132 | |
NM_114847.5 | NP_190556.2 | 5 - 6 | 700 | 132 | ||
GenBank mRNA | AY072429.1 | - | 5 - 6 | 554 | 132 | |
AY114718.1 | - | 5 - 6 | 436 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |