Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLP3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339487.1 | NP_001325881.1 | 2 - 3 | 357 | 89 | |
NM_114956.3 | NP_190665.2 | 3 - 4 | 288 | 89 | ||
GenBank mRNA | AK229066.1 | - | 3 - 4 | 244 | 89 | |
BT010901.1 | - | 2 - 3 | 155 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |