Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGRL1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203773.1 | NP_001190702.1 | 10 - 11 | 1009 | 79 | |
NM_117266.4 | NP_192933.2 | 9 - 10 | 1046 | 79 | ||
GenBank mRNA | AK117721.1 | - | 9 - 10 | 916 | 79 | |
BT005305.1 | - | 9 - 10 | 867 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |