Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_122022.3 | NP_197515.1 | 2 - 3 | 540 | 163 | |
GenBank mRNA | AY075605.1 | - | 2 - 3 | 435 | 163 | |
BT000868.1 | - | 2 - 3 | 393 | 163 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |