Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343647.1 | NP_001330416.1 | 1 - 2 | 468 | 126 | |
NM_122031.4 | NP_197524.1 | 1 - 2 | 551 | 126 | ||
GenBank mRNA | D30807.1 | - | 1 - 2 | 275 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Arabidopsis thaliana Probe spans exons |