Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036838.2 | NP_001031915.1 | 13 - 14 | 2677 | 101 | |
NM_001343687.1 | NP_001332603.1 | 13 - 14 | 2477 | 101 | ||
NM_122090.4 | NP_197583.1 | 12 - 13 | 3076 | 101 | ||
GenBank mRNA | AK316826.1 | - | 12 - 13 | 2483 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |