Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B73 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343692.1 | NP_001332397.1 | 2 - 3 | 433 | 57 | |
NM_001343693.1 | NP_001332398.1 | 1 - 2 | 183 | 57 | ||
NM_122108.4 | NP_197599.1 | 2 - 3 | 443 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |