Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_122138.3 | NP_197625.1 | 10 - 11 | 1427 | 87 | |
GenBank mRNA | AY084606.1 | - | 10 - 11 | 1341 | 87 | |
BT000923.1 | - | 10 - 11 | 1218 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |